missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARHGEF9 Partial ORF (NP_056000.1, 419 a.a. - 516 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_056000.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23229 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124516
|
Abnova™
H00023229-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134516
|
Abnova™
H00023229-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ARHGEF9 belongs to a family of Rho-like GTPases that act as molecular switches by cycling from the active GTP-bound state to the inactive GDP-bound state. These proteins are key regulators of the actin cytoskeleton and are involved in cell signaling.[supplied by OMIM]
Sequence: AFREERKMVQEDEKIGFEISENQKRQAAMTVRKVPKQKGVNSARSVPPSYPPPQDPLNHGQYLVPDGIAQSQVFEFTEPKRSQSPFWQNFSRLTPFKKSpecifications
NP_056000.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COLLYBISTIN/HPEM-2/KIAA0424/PEM-2/PEM2 | |
ARHGEF9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23229 | |
ARHGEF9 (Human) Recombinant Protein (Q01) | |
AFREERKMVQEDEKIGFEISENQKRQAAMTVRKVPKQKGVNSARSVPPSYPPPQDPLNHGQYLVPDGIAQSQVFEFTEPKRSQSPFWQNFSRLTPFKK | |
RUO | |
ARHGEF9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |