missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARHGEF6 Partial ORF (AAH39856.1, 61 a.a. - 175 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAH39856.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9459 |
Molecular Weight (g/mol) | 38.06kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180716
|
Abnova™
H00009459-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16170716
|
Abnova™
H00009459-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein belongs to a family of cytoplasmic proteins that activate the Ras-like family of Rho proteins by exchanging bound GDP for GTP. It may form a complex with G proteins and stimulate Rho-dependent signals. This protein is activated by PI3-kinase. Mutations in this gene can cause X-chromosomal non-specific mental retardation. [provided by RefSeq]
Sequence: QTEADCINNINDFLKGCATLQVEIFDPDDLYSGVNFSKVLSTLLAVNKATEDQLSERPCGRSSSLSAANTSQTNPQGAVSSTVSGLQRQSKTVEMTENGSHQLIVKARFNFKQTNSpecifications
AAH39856.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.06kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COOL2/Cool-2/KIAA0006/MRX46/PIXA/alpha-PIX/alphaPIX | |
ARHGEF6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9459 | |
ARHGEF6 (Human) Recombinant Protein (Q01) | |
QTEADCINNINDFLKGCATLQVEIFDPDDLYSGVNFSKVLSTLLAVNKATEDQLSERPCGRSSSLSAANTSQTNPQGAVSSTVSGLQRQSKTVEMTENGSHQLIVKARFNFKQTN | |
RUO | |
ARHGEF6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |