missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Human Apolipoprotein M/ApoM (NP_061974) VersaClone cDNA
Klik for at se tilgængelige muligheder
Mængde:
10 μg
Beskrivelse
The Human Apolipoprotein M/ApoM (NP_061974) VersaClone cDNA clone contains the complete ORF for Apolipoprotein M/ApoM, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Apolipoprotein M/ApoM insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Tekniske data
Tekniske data
| Gene | hAPOM |
| Adgangsnummer | NP_061974 |
| Koncentration | 10 μg at 0.2 μg/μL |
| Formulering | cDNA is provided in 10 mM Tris-Cl |
| Indsæt type | 580bp |
| pH | 8.5 |
| Gen-id (Entrez) | 55937 |
| Gene Alias | apolipoprotein M, ApoM, Apo-M, G3A, HSPC336, MGC22400, NG20, NG20-like protein, Protein G3a |
| Indhold og opbevaring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Arter | Human |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion