Learn More
Gibco™ Human ANG-1 Recombinant Protein, PeproTech®
Description
130-06-1MG will be provided as 10 x 100 μg (130-06-100UG). 130-06-500UG will be provided as 5 x 100 μg (130-06-100UG). Recombinant Human ANG-1, derived from HeLa cells, is a C-terminal histidine tagged glycoprotein which migrates with an apparent molecular mass of 60.0-70.0 kDa by SDS-PAGE under reducing conditions. Sequencing analysis shows N-terminal sequences starting with Ser-20 and with Asp-70 of the 498 amino acid precursor protein. The calculated molecular weight of Recombinant Human ANG-1 is 56.3 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Specifications
Specifications
Accession Number | Q15389 |
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 284 |
Molecular Weight (g/mol) | 56.3 |
Name | Human ANG-1 |
Quantity | 5 x 100 μg |
Source | HeLa cells |
Storage Requirements | -20°C |
Regulatory Status | RUO |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.