missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ALG12 Partial ORF (NP_077010, 369 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079087-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16169926

  • 2665.00 DKK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the glycosyltransferase 22 family. The encoded protein catalyzes the addition of the eighth mannose residue in an alpha-1,6 linkage onto the dolichol-PP-oligosaccharide precursor (dolichol-PP-Man(7)GlcNAc(2)) required for protein glycosylation. Mutations in this gene have been associated with congenital disorder of glycosylation type Ig (CDG-Ig)characterized by abnormal N-glycosylation. [provided by RefSeq]

Sequence: NYPGGVAMQRLHQLVPPQTDVLLHIDVAAAQTGVSRFLQVNSAWRYDKREDVQPGTG
Specifications

Specifications

NP_077010
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.01kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NYPGGVAMQRLHQLVPPQTDVLLHIDVAAAQTGVSRFLQVNSAWRYDKREDVQPGTG
RUO
ALG12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
79087
ALG12 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ECM39/MGC111358/MGC3136/PP14673/hALG12
ALG12
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers