Learn More
Abnova™ Human ADIPOR1 Partial ORF (NP_057083.2, 72 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051094-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
The adiponectin receptors, ADIPOR1 and ADIPOR2 (MIM 607946), serve as receptors for globular and full-length adiponectin (MIM 605441) and mediate increased AMPK (see MIM 602739) and PPAR-alpha (PPARA; MIM 170998) ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin (Yamauchi et al., 2003 [PubMed 12802337]).[supplied by OMIM]
Sequence: QAHHAMEKMEEFVYKVWEGRWRVIPYDVLPDWLKDNDYLLHGHRPPMPSFRACFKSIFRIHTETGSpecifications
NP_057083.2 | |
Liquid | |
51094 | |
ADIPOR1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACDCR1/CGI-45/CGI45/FLJ25385/FLJ42464/PAQR1/TESBP1A | |
ADIPOR1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QAHHAMEKMEEFVYKVWEGRWRVIPYDVLPDWLKDNDYLLHGHRPPMPSFRACFKSIFRIHTETG | |
RUO | |
ADIPOR1 | |
Wheat Germ (in vitro) | |
GST |