missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADAMTS6 Partial ORF (NP_922932.2, 725 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011174-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133836

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. [provided by RefSeq]

Sequence: DSLPRGGYMEVVQIPRGSVHIEVREVAMSKNYIALKSEGDDYYINGAWTIDWPRKFDVAGTAFHYKRPTDEPESLEALGPTSENLIVMVLLQEQNLGIRY
Specifications

Specifications

NP_922932.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DSLPRGGYMEVVQIPRGSVHIEVREVAMSKNYIALKSEGDDYYINGAWTIDWPRKFDVAGTAFHYKRPTDEPESLEALGPTSENLIVMVLLQEQNLGIRY
RUO
ADAMTS6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11174
ADAMTS6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADAM-TS6
ADAMTS6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers