Learn More
Abnova™ Human ADAMTS3 Partial ORF (NP_055058.1, 1048 a.a. - 1128 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009508-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene is the major procollagen II N-propeptidase. A deficiency of this protein may be responsible for dermatosparaxis, a genetic defect of connective tissues. [provided by RefSeq]
Sequence: ESCSKRSSTLPPPYLLEAAETHDDVISNPSDLPRSLVMPTSLVPYHSETPAKKMSLSSISSVGGPNAYAAFRPNSKPDGANSpecifications
NP_055058.1 | |
Liquid | |
9508 | |
ADAMTS3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADAMTS-4/KIAA0366 | |
ADAMTS3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.65kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ESCSKRSSTLPPPYLLEAAETHDDVISNPSDLPRSLVMPTSLVPYHSETPAKKMSLSSISSVGGPNAYAAFRPNSKPDGAN | |
RUO | |
ADAMTS3 | |
Wheat Germ (in vitro) | |
GST |