missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human ADAM33 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5565
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ADAM33 Monoclonal specifically detects ADAM33 in Human samples. It is validated for Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Tekniske data
| ADAM33 | |
| Monoclonal | |
| LYOPH | |
| Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9BZ11 | |
| ADAM33 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM33 Val30-Leu501 (predicted) Accession # Q9BZ11 | |
| 100 μg | |
| Primary | |
| Detects human ADAM33 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse ADAM33 is observed. | |
| Human | |
| Purified |
| Immunoprecipitation, Flow Cytometry, CyTOF | |
| 508101 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, ADAM 33, ADAM metallopeptidase domain 33, C20orf153, chromosome 20 open reading frame 153, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein, DJ964F7.1, DKFZp434K0521, EC 3.4.24.-, FLJ35308, FLJ36751, MGC149823, MGC71889 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 80332 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion