missing translation for 'onlineSavingsMsg'
Få mere at vide

Human ADAM12 Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. p-7062277
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18747137 25 μg
18082102 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18747137 Leverandør R&D Systems Leverandørnr. AF4416SP

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Sheep Polyclonal Antibody

ADAM12 Polyclonal specifically detects ADAM12 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ADAM12
Applikationer Western Blot, Immunoprecipitation
Klassifikation Polyclonal
Koncentration LYOPH
Konjugeret Unconjugated
Fortynding Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Formulering Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Genadgangsnr. AAC08702
Gene Alias a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin alpha, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.-
Gensymboler ADAM12
Værtsarter Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAM12 Arg29-Ser513 Accession # AAC08702
Oprensningsmetode Affinity Purified
Mængde 25 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 8038
Testspecificitet Detects human ADAM12 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) ADAM8, rhADAM19, and recombinant mouse ADAM33 is observed.
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Målarter Human
Indhold og opbevaring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.