Learn More
Abnova™ Human ACBD3 Partial ORF (NP_073572, 73 a.a. - 171 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064746-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is involved in the maintenance of Golgi structure and function through its interaction with the integral membrane protein giantin. It may also be involved in the hormonal regulation of steroid formation. [provided by RefSeq]
Sequence: RRLEQRWGFGLEELYGLALRFFKEKDGKAFHPTYEEKLKLVALHKQVLMGPYNPDTCPEVGFFDVLGNDRRREWAALGNMSKEDAMVEFVKLLNRCCHLSpecifications
NP_073572 | |
Liquid | |
64746 | |
ACBD3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GCP60/GOCAP1/GOLPH1/PAP7 | |
ACBD3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RRLEQRWGFGLEELYGLALRFFKEKDGKAFHPTYEEKLKLVALHKQVLMGPYNPDTCPEVGFFDVLGNDRRREWAALGNMSKEDAMVEFVKLLNRCCHL | |
RUO | |
ACBD3 | |
Wheat Germ (in vitro) | |
GST |