Learn More
Abnova™ Human ACADS Full-length ORF (AAH25963.1, 1 a.a. - 412 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000035-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a a tetrameric mitochondrial flavoprotein, which is a member of the acyl-CoA dehydrogenase family. This enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Mutations in this gene have been associated with Short Chain Acyl-CoA Dehydrogenase Deficiency. [provided by RefSeq]
Sequence: MAAALLARASGPARRALCPRAWRQLHTIYQSVELPETHQMLLQTCRDFAEKELFPIAAQVDKEHLFPAAQVKKMGGLGLLAMDVPEELGGAGLDYLAYAIAMEEISRGCASTGVIMSVNNSLYLGPILKFGSKEQKQAWVTPFTSGDKIGCFALSEPGNGSDAGAASTTARAEGDSWVLNGTKAWITNAWEASAAVVFASTDRALQNKSISAFLVPMPTPGLTLGKKEDKLGIRGSSTANLIFEDCRIPKDSILGEPGMGFKIAMQTLDMGRIGIASQALGIAQTALDCAVNYAENRMAFGAPLTKLQVIQFKLADMALALESARLLTWRAAMLKDNKKPFIKEAAMAKLAASEAATAISHQAIQILGGMGYVTEMPAERHYRDARITEIYEGTSEIQRLVIAGHLLRSYRSSpecifications
AAH25963.1 | |
Liquid | |
35 | |
ACADS (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACAD3/SCAD | |
ACADS | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ACADS | |
Wheat Germ (in vitro) | |
GST |