missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AAMP Partial ORF (NP_001078.2, 111 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_001078.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 14 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073585
|
Abnova™
H00000014-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083585
|
Abnova™
H00000014-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The gene product is an immunoglobulin-type protein. It is found to be expressed strongly in endothelial cells, cytotrophoblasts, and poorly differentiated colon adenocarcinoma cells found in lymphatics. The protein contains a heparin-binding domain and mediates heparin-sensitive cell adhesion. [provided by RefSeq]
Sequence: GEDDKAFVWRLSDGELLFECAGHKDSVTCAGFSHDSTLVATGDMSGLLKVWQVDTKEEVWSFEAGDLEWMEWHPRAPVLLAGTADGNTWMWKVPNGDCKTSpecifications
NP_001078.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AAMP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
14 | |
AAMP (Human) Recombinant Protein (Q01) | |
GEDDKAFVWRLSDGELLFECAGHKDSVTCAGFSHDSTLVATGDMSGLLKVWQVDTKEEVWSFEAGDLEWMEWHPRAPVLLAGTADGNTWMWKVPNGDCKT | |
RUO | |
AAMP | |
Recombinant | |
wheat germ expression system |