Learn More
Hamilton™ Small Hub Removable Needle, Custom Length
Suitable for use with removable (RN)-style syringes thar are 100μL or smaller. Hamilton™ Small Hub Removable Needle, Custom Length is available in lengths between 0.375 and 12 inches.
Brand: Hamilton™ 7803-01
2934.73 DKK valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Autoclavable
- Gas sterilizable
- 6/Pk
Specifications
Small Hub Needle | |
27 G | |
0.375 to 12 in. | |
Custom Point Style (2, 3, or 4) |