Få mere at vide
Tocris Bioscience™ HTH 01-015
Potent and selective NUAK1 inhibitor
Brand: Tocris Bioscience™ 5622/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
HTH 01-015 is a potent and selective NUAK1 inhibitor (IC50 = 100 nM). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells in vitro, to a similar extent as NUAK1 knockdown models.Tekniske data
| 5,13-Dihydro-4,5,13-trimethyl-2-[[1-(4-piperidinyl)-1H-pyrazol-4-yl]amino]-6H-naphtho[2,3-e]pyrimido[5,4-b][1,4]diazepin-6-one | |
| 1613724-42-7 | |
| 50 mg |