Få mere at vide
BOLA HT Replacement Inner Parts
Trådstørrelse:
32
45
Diameter (metrisk):
3 mm
3,2 mm
5 mm
6 mm
8 mm
9,5 mm
9,52 mm
12,7 mm
14 mm
16 mm
18 mm
18,01 mm
19,05 mm
20 mm
21 mm
22 mm
30 mm
32 mm
Beskrivelse
Consisting of a v-ring (PPS), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm (not exposed to the medium). Good chemical resistance, the medium is only exposed to PTFE.
Tekniske data
Tekniske data
| Certificeringer/compliance | FDA |
| Diameter (metrisk) | 9,5 mm |
| Autoklaverbar | Autoclavable |
| Trådstørrelse | 32 |
| Farve | White |
| Fleksibilitet | Rigid |