Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup
HT ExoSAP-IT Fast High-Throughput PCR Product Clean-up, with its shortened 14 minute protocol, is a highly stable, customized formulation of the original ExoSAP-IT reagent
3120.00 DKK - 35250.00 DKK
Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15839916
View Documents Promotion Details
Applied Biosystems™
785951EA
480 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15849916
View Documents Promotion Details
Applied Biosystems™
785951PK
5760 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • 14 minute protocol is half the time of standard protocols
  • One-tube PCR clean-up - add HT ExoSAP-IT Fast reagent directly to PCR product
  • 100% sample recovery - no loss of PCR products regardless of the fragment size
  • Stable at 4°C for one month
  • Removes excess primers and dNTPs - does not interfere with downstream applications sequencing, SNP analysis, single base extension, fragment analysis
  • It is ideal for high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
  • Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design