Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup
HT ExoSAP-IT Fast High-Throughput PCR Product Clean-up, with its shortened 14 minute protocol, is a highly stable, customized formulation of the original ExoSAP-IT reagent
3120.00 DKK - 35250.00 DKK
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15839916
|
Applied Biosystems™
785951EA |
480 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15849916
|
Applied Biosystems™
785951PK |
5760 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- 14 minute protocol is half the time of standard protocols
- One-tube PCR clean-up - add HT ExoSAP-IT Fast reagent directly to PCR product
- 100% sample recovery - no loss of PCR products regardless of the fragment size
- Stable at 4°C for one month
- Removes excess primers and dNTPs - does not interfere with downstream applications sequencing, SNP analysis, single base extension, fragment analysis
- It is ideal for high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
- Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Order Info
Each
For Research Use Only. Not for use in diagnostic procedures.