Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up
HT ExoSAP-IT Fast High-Throughput PCR Product Clean-up, with its shortened 14 minute protocol, is a highly stable, customized formulation of the original ExoSAP-IT reagent
Brand: Applied Biosystems™ 7859520RXN
Additional Details : Weight : 0.01000kg
Description
- 14 minute protocol is half the time of standard protocols
- One-tube PCR clean-up - add HT ExoSAP-IT Fast reagent directly to PCR product
- 100% sample recovery - no loss of PCR products regardless of the fragment size
- Stable at 4°C for one month
- Removes excess primers and dNTPs - does not interfere with downstream applications sequencing, SNP analysis, single base extension, fragment analysis
- It is ideal for high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
- Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Order Info
Each
Specifications
Fast High-Throughput PCR Product Clean-up, 20 rxns, Highly Stable, 4 Minute Protocol, Ideal for High-throughput PCR Product Clean-up Using Liquid Handling Platforms and Multi-Channel Pipettes | |
20 rxns | |
1000 Reactions |
For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes | |
HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up | |
–20°C |
For research use only