missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ HSP27 Recombinant Rabbit Monoclonal Antibody (SAIC-23B-22)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547899
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Specificity: Recognises human HSPB1.
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock or stress proteins. HSPB1 (heat shock protein beta-1 or HSP27) is a small heat shock protein which functions as a molecular chaperone that maintains denatured proteins in a folding-competent state. It plays a role in stress resistance and actin organization. Through its molecular chaperone activity, HSP27 regulates numerous biological processes including the phosphorylation and the axonal transport of neurofilament proteins. Mutations in the gene can result in Charcot-Marie-Tooth disease 2F and Neuronopathy distal hereditary motor 2B.
Tekniske data
| HSP27 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P04792 | |
| HSPB1 | |
| Peptide DGVVEITGK derived from Heat shock protein beta-1, involved in stress resistance and actin organisation, conjugated to KLH. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
| SAIC-23B-22 | |
| Unconjugated | |
| HSPB1 | |
| 27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3315 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion