missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
HSF1 Antibody (009), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90081DL594
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
HSF1 Monoclonal specifically detects HSF1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Tekniske data
| HSF1 | |
| Monoclonal | |
| DyLight 594 | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human HSF1 (Q00613-1; Asp2-Ser529). | |
| 0.1 mL | |
| Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
| 3297 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 9 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Spot an opportunity for improvement?Share a Content Correction