Learn More
Abcam HRV14 3C (PreScission) Protease Agarose Beads
Product Code. 32115515 Shop All Abcam Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity Unit Size
32115515 5 mL 5mL
1 options
Product Code. 32115515 Supplier Abcam Supplier No. ab2868835mL
 Restricted Item
Forventet forsendelse: 01-12-2026
HRV14 3C (PreScission) Protease Agarose Beads, ab286883, (previously known as #M1029) is prepared by covalent coupling active HRV14 3C Protease to activated 4% cross-linked agarose beads.
HRV14 3C (PreScission) Protease Agarose Beads, ab286883, (previously known as #M1029) is prepared by covalent coupling active HRV14 3C Protease to activated 4% cross-linked agarose beads.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 5 mL
Regulatory Status RUO