Learn More
Abcam HRP Conjugate Stabilizer - LifeXtend
Product Code. 32113393 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. Quantity Unit Size
32113393 1000 mL 1000mL
1 options
Product Code. 32113393 Supplier Abcam Supplier No. ab2705481000mL
 Restricted Item
Forventet forsendelse: 24-11-2026
HRP Conjugate Stabilizer - LifeXtend (ab270548) is a proprietary multi-component reagent system that protects antibody-HRP conjugates from instability, microbrial attack, and denaturation, thus ensuring the best possible performance in experiments performed at room temperature.
HRP Conjugate Stabilizer - LifeXtend (ab270548) is a proprietary multi-component reagent system that protects antibody-HRP conjugates from instability, microbrial attack, and denaturation, thus ensuring the best possible performance in experiments performed at room temperature.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1000 mL
Regulatory Status RUO