missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
HRAS Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99921C
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
HRAS Polyclonal specifically detects HRAS in Human samples. It is validated for Western Blot, ELISA.
Tekniske data
| HRAS | |
| Polyclonal | |
| Western Blot, ELISA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Produced in rabbits immunized with purified, recombinant HRAS (P01112; Met1-Cys186). | |
| 0.1 mL | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 3265 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion