Få mere at vide
Hoveyda-Grubbs Catalyst 1st Generation, TRC
Artikelnummer. 30400207
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30400207 500 mg
30402829 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30400207 Leverandør Toronto Research Chemicals Leverandørnr. TRCH700520500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 203714-71-0
Molekylær formel C28 H45 Cl2 O P Ru
Molekylvægt (g/mol) 600.61
Synonym Hoveyda-Grubbs Catalyst 1st Generation, (SP-5-41)-Dichloro[[2-(1-methylethoxy-kO)phenyl]methylene-kC](tricyclohexylphosphine)ruthenium
IUPAC navn 8,8-dichloro-7-propan-2-yl-8-(tricyclohexyl-λ^{4}-phosphanyl)-7λ^{3}-oxa-8λ^{6}-ruthenabicyclo[4.3.0]nona-1,3,5,8-tetraene
SMIL CC(C)[O]1c2ccccc2C=[Ru]1(Cl)(Cl)[P](C3CCCCC3)(C4CCCCC4)C5CCCCC5
Tekniske data
Kemisk navn eller materiale Dichloro((2-(1-methylethoxy-alpha-O)phenyl)methylene-alpha-C)(tricyclohexylphosphine)ruthenium
Mængde 500 mg
InChI formel InChI=1S/C18H33P.C10H12O.2ClH.Ru/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-8(2)11-10-7-5-4-6-9(10)3;;;/h16-18H,1-15H2;3-8H,1-2H3;2*1H;/q;;;;+2/p-2
Formel vægt 600.1629