Få mere at vide
Testo™ 425 Compact Thermal Anemometer
Artikelnummer. 10084632
Højde (metrisk):
182 mm
Artikelnummer. 10084632
Brand: Testo™ 0560 4251
Use this anemometer for a variety of applications. Testoterm™ 425 Compact Thermal Anemometer measures the flow speed and automatically calculates the volumetric flow during the testing of ventilation ducts and air conditioning systems.
  • Temperature and flow measurement as well as volumetric flow calculation
  • Permanently attached extendible telescopic probe maximum length 820mm, diameter 7.5mm
  • With single and multi-point averaging
  • Hold function, display of min/max value, Auto-off function
Tekniske data
Type Hot Wire Anemometer
Højde (metrisk) 182 mm
Længde (metrisk) 64 mm
Temperatur (metrisk) 20°C to 50°C
Vægt (metrisk) 285 g