missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Host Cell Factor 1/HCFC1 Antibody [PE], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals AF6254P
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Host Cell Factor 1/HCFC1 Polyclonal antibody specifically detects Host Cell Factor 1/HCFC1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
Tekniske data
| Host Cell Factor 1/HCFC1 | |
| Polyclonal | |
| PBS | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| PE | |
| CFFC1 factor, HCF, HCF1HFC1, HCF-1VP16 accessory protein, host cell factor C1 (VP16-accessory protein), MGC70925, VCAFhost cell factor 1 | |
| E. coli-derived recombinant human Host Cell Factor 1/HCFC1, Ala1626-Val1836, Accession # P51610 | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Endocrinology | |
| 3054 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion