Få mere at vide
Honeywell™ Horizon Safety Spectacles
:
Clear
Beskrivelse
- Adjustable temple wires
- Flip-up window
- Durable polyamide frame
- Molded sideshields
- Can be equipped with 4 types of flip-up windows depending on application
- Frame designed to accept prescription lenses
- Easily adjustable temples
- Integrated sub-orbital protection
Tekniske data
Tekniske data
| Linsebelægning | Scratch-resistant |
| Certificeringer/overholdelse (Europa) | EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: C1570.1HON, EC Attestation: C1570.1HON, EN C1570.1HON |
| Linsefarve | Clear |
| Til brug med (applikation) | General use and protection against impact (45m/s), mechanical work: grinding, turning, milling, UV protection, welding applications with the welding flip-up, can be fitted with prescription lenses |
| Ramme farve | Clear |
| Linse materiale | Polycarbonate |
| Rammemateriale | Grilamid |
| UV beskyttelse | 0.999 |