missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medicom™ Hopen™ Zipper-Closure Hygiene Gown
Medicom™ Hopen™ Zipper-Closure Hygiene Gown with no pockets, suitable for multiple uses
Brand: Medicom™ 40.635/04
Additional Details : Weight : 4.94000kg
Description
- This Zipper-Closure Hygiene Gown protects the operator from work-related contamination
- Also protects against sensitive products and processes that could be contaminated in a protected atmosphere
- Pleasant and light, polypropylene fabric is breathable
- Individually packaged for controlled hygiene and distribution
- Without pockets to prevent falling objects
Specifications
X-Large | |
Polypropylene | |
Zipper | |
No | |
Radioactive Particles, Infectious Agents | |
Sewn | |
Category 1 Personal Protective Equipment |
Unisex | |
Blue | |
Elastic | |
Non-sterile | |
None | |
Manufactured under an ISO 9001 Certified Quality System | |
Individual, Bag |