missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Spacel™ 3000 RA/EBJ Coveralls with Hood

Catalog Number p-8011031
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Spacel™ 3000 RA/EBJ Coveralls are made from three-layer polyethylene film with good abrasion, tear, and perforation resistance. With an elasticated hood, wrists, and ankles plus sealed seams and a sealed zipper/adhesive flap closure, the liquid-tight design protects workers from chemical hazards.

  • Product Type: Coveralls
  • Material: Polyethylene
  • Gender: Unisex
  • Color: Yellow
  • Disposable: Yes
  • Sterility: Non-Sterile
  • Resistance: Chemical, Abrasion, Tear, and Perforation
  • Certification: EC Category 3, Protection Level: 3, 4, 5, 6b
  • Hood, Wrist, and Ankle Type: Elasticated
  • Closure Type: Sealed Zipper/Double Adhesive Flap
  • Seam Type: Sealed
  • For Use With: Chemistry, Petrochemistry, In-Shore/Off-Shore Oil Industry, Industrial Maintenance, Pharmaceutical Industry, Spoiled Beach Cleaning, Transportation of Hazardous Materials, Industrial Sub-Contracting Cleaning, Sanitation, Waste Processing, Emergency Services
TRUSTED_SUSTAINABILITY
Product Type Hooded Coveralls
Size Small
Gender Unisex
Material Polyethylene
Color Yellow
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Closure Type Sealed Zipper with Double-adhesive Flap
Seam Type Sealed
Sterility Non-sterile
Protection Class Protection Level: 3, 4, 5, 6b
Resistance Chemicals, Abrasion, Tears, Perforation
Construction Three-Layer Film
Certifications/Compliance (Europe) EC Category: 3
Thickness 100 μm
For Use With (Application) Chemistry, Petrochemistry, In-Shore/Off-Shore Oil Industry, Industrial Maintenance, Pharmaceutical Industry, Spoiled Beach Cleaning, Transportation of Hazardous Materials, Industrial Sub-Contracting Cleaning, Sanitation, Waste Processing, Emergency Services
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'