Learn More
Sperian™ Honeywell™ Bionic™ FogBan™ Anti-Scratch Clear Faceshields
Product Code. 11836950
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
11836950 Polycarbonate Each
1 options
Product Code. 11836950 Supplier Sperian™ Supplier No. 1011624
  • ONLINE EXCLUSIVE PRICE
    790.66 DKK
    443.72 DKK / Pakke med 1
    Spare 346.94 DKK
    43% Slukket
View all promotions
Forventet forsendelse: 28-12-2026
Provides the most comfortable fit in the industry through its many adjustable features. Sperian™ Honeywell™ Bionic™ FogBan™ Anti-Scratch Clear Faceshields offer maximum protection.
  • Extended top of head and chin protection
  • Large superior quality visors for maximum visibility
  • Easily replaced visors available in different materials, tints and finishes to better suit individual needs
  • Comfortable fit with goggles or respirators
  • Locking mechanism for more complete safety
  • Dielectric design
  • Fully adjustable ratchet headgear
  • Fully adjustable headband
  • Multiple visor tilt positions
  • Extra-soft cushioning on all parts of the shield in contact with the head
  • Breathable extra-soft sweatband is removable for easy washing and replacement
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bionic Faceshield
No. per Pack 1 Ea.
Lens Material Polycarbonate