Få mere at vide
Homovanillic Acid-d5, TRC
Artikelnummer. 30404282
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30404282 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404282 Leverandør Toronto Research Chemicals Leverandørnr. TRCH6695061MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Homovanillic Acid-d5
CAS 53587-32-9
Renhedsgrad noter HPLC
Molekylær formel C9 D5 H5 O4
Mængde 1 mg
Synonym 4-Hydroxy-5-methoxy-benzene-2,3,6-d3-acetic-α,α-d2 Acid, (4-Hydroxy-3-methoxyphenyl-2,5,6-d3)acetic-2,2-d2 Acid, Benzene-2,3,6-d3-acetic-α,α-d2 acid, 4-hydroxy-5-methoxy- (9CI), 2-[4-Hydroxy-3-(trideuteriomethoxy)phenyl]dideuterioacetic acid, 4-Hydroxy-3-(methoxy-d3)benzeneacetic-d2 acid, 2-[4-Hydroxy-3-(methoxy-d3)phenyl]acetic-2,2-d2 acid, 3-(Methoxy-d3)-4-hydroxyphenylacetic-d2 acid, 3'-(Methoxy-d3)-4'-hydroxyphenylacetic-2,2-d2 acid, Homovanillic acid D5, Vanilacetic acid-d5, 4-Hydroxy-3-methoxyphenyl-D3-acetic-D2 acid, HVA-D5
InChI formel InChI=1S/C9H10O4/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4,10H,5H2,1H3,(H,11,12)/i2D,3D,4D,5D2
SMIL [2H]c1c([2H])c(c([2H])c(OC)c1O)C([2H])([2H])C(=O)O
IUPAC navn 2,2-dideuterio-2-(2,3,6-trideuterio-4-hydroxy-5-methoxyphenyl)acetic acid
Molekylvægt (g/mol) 187.2041
Formel vægt 187.0893
Procent renhed >95