Få mere at vide
Homochlorcyclizine Dihydrochloride, LoGiCal
Artikelnummer. 30458918
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30458918 5 mg
1 options
Artikelnummer. 30458918 Leverandør LoGiCal Leverandørnr. LGCFOR3101.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Homochlorcyclizine Dihydrochloride
CAS 1982-36-1
Analyt- eller komponentnavne Homochlorcyclizine Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C19 H23 Cl N2 . 2 Cl H
Certificeringer/compliance ISO 9001
Synonym Homochlorcyclizine Dihydrochloride, 1-[(4-Chlorophenyl)phenylmethyl]hexahydro-4-methyl-1H-1,4-diazepine hydrochloride (1:2), 1-(p-Chloro-α-phenylbenzyl)hexahydro-4-methyl-1H-1,4-diazepine dihydrochloride, 1-[(4-Chlorophenyl)phenylmethyl]hexahydro-4-methyl-1H-1,4-diazepine dihydrochloride, 1-((4-Chlorophenyl)(phenyl)methyl)-4-methyl-1,4-diazacycloheptane dihydrochloride, Homochlorcyclizine dihydrochloride, Homochlorcyclizine hydrochloride, NSC 169498, SA 97 dihydrochloride
InChI formel InChI=1S/C19H23ClN2.2ClH/c1-21-12-5-13-22(15-14-21)19(16-6-3-2-4-7-16)17-8-10-18(20)11-9-17;;/h2-4,6-11,19H,5,12-15H2,1H3;2*1H
Løsningstype Neat
SMIL Cl.Cl.CN1CCCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC navn 1-[(4-chlorophenyl)-phenylmethyl]-4-methyl-1,4-diazepane;dihydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 387.77
Mængde 5 mg
Formel vægt 386.1083 g/mol