Få mere at vide
Homo-PROTAC cereblon degrader 1, MedChemExpress
Artikelnummer. 16467217
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16467217 5 mg
16444498 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16467217 Leverandør Medchem Express Leverandørnr. HY111594
MedChemExpress Homo-PROTAC cereblon degrader 1 (compound 15a) is a highly potent and efficient Cereblon (CRBN) degrader with only minimal effects on IKZF1 and IKZF3.
Homo-PROTAC cereblon degrader 1 (compound 15a) is a highly potent and efficient Cereblon (CRBN) degrader with only minimal effects on IKZF1 and IKZF3.
Tekniske data
Kemisk navn eller materiale Homo-PROTAC cereblon degrader 1
CAS 2244520-98-5
Farve Green
Renhedsgrad noter Research
Molekylær formel C32H32N6O10
Mængde 5 mg
Opløselighedsinformation DMSO : 200 mg/mL (302.74 mM; Need ultrasonic)
SMIL O=C(N1C2CCC(NC2=O)=O)C3=CC=CC(NCCOCCOCCNC4=C5C(C(N(C6CCC(NC6=O)=O)C5=O)=O)=CC=C4)=C3C1=O
Molekylvægt (g/mol) 660.63
Formel vægt 660.63
Procent renhed 98.31%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death