Få mere at vide
Homatropine Methylbromide, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Homatropine methylbromide |
| CAS | 80-49-9 |
| Analyt- eller komponentnavne | Homatropine methylbromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H24 N O3 . Br |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Homatropine Methylbromide, (1R,3r,5S)-3-[[(2RS)-2-Hydroxy-2-phenylacetyl]oxy]-8,8-dimethyl-8-azoniabicyclo[3.2.1]octane bromide |
| InChI formel | InChI=1S/C17H24NO3.BrH/c1-18(2)13-8-9-14(18)11-15(10-13)21-17(20)16(19)12-6-4-3-5-7-12;/h3-7,13-16,19H,8-11H2,1-2H3;1H/q+1;/p-1/t13-,14+,15+,16-;/m1./s1 |
| Løsningstype | Neat |
| SMIL | [Br-].C[N+]1(C)[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](O)c3ccccc3 |
| Vis mere |