Få mere at vide
Homatropine Hydrobromide, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Homatropine Hydrobromide |
| CAS | 51-56-9 |
| Analyt- eller komponentnavne | Homatropine Hydrobromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H21 N O3 . Br H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Homatropine hydrobromide, (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl (2RS)-2-hydroxy-2-phenylacetate hydrobromide, Homatropine bromide, Isopto Homatropine, Mandelic acid tropine ester hydrobromide, 1alphaH,5alphaH-Tropan-3alpha-ol, mandelate (ester), hydrobromide (8CI), Benzeneacetic acid, alpha-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide (9CI), Benzeneacetic acid, alpha-hydroxy-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide, endo-, (+/-)-Homatropine bromide, Benzeneacetic acid, alpha-hydroxy-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, hydrobromide (1:1) |
| InChI formel | InChI=1S/C16H21NO3.BrH/c1-17-12-7-8-13(17)10-14(9-12)20-16(19)15(18)11-5-3-2-4-6-11;/h2-6,12-15,18H,7-10H2,1H3;1H/t12-,13+,14+,15-;/m1./s1 |
| Løsningstype | Neat |
| SMIL | Br.CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](O)c3ccccc3 |
| Vis mere |