Learn More
Hoefer™ HE-PLUS Casting Kits
For use with HE-PLUS Horizontal Electrophoresis System
Brand:  Hoefer™ HE-PLUS18
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15378693
  • 1652.17 DKK / Each
Estimated Shipment
18-06-2021
Specifications
HE-plus Horizontal Electrophoresis System
Standard Casting Kit
12.5 x 13 cm
One large gel tray (12.5 x 13cm), two medium gel trays (12.5 x 6cm), four combs (14/28-well) and casting stand