Få mere at vide
MedChemExpress Hoechst S 769121
Tekniske data
Tekniske data
| Indhold og opbevaring | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Mængde | 5 mg |
| Forsendelsestilstand | Room temperature |
| Opløselighed | H2O : ≥ 6 mg/mL (10.05 mM) |
| Molekylvægt (g/mol) | 596.96 g/mol |
| CAS | 74681-68-8 |
| Synonym | Nuclear yellow |