Få mere at vide
hnRNP M Polyclonal Antibody
hnRNP M Polyclonal Antibody
Brand: Invitrogen PA5121046
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
hnRNP M Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Tekniske data
| hnRNP M | |
| Polyclonal | |
| Unconjugated | |
| HNRNPM | |
| 2610023M21Rik; AA409009; CEA receptor; CEAR; heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; HNRNPM; HNRNPM4; Hnrpm; Hnrpm4; HTGR1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein M; M4 protein; mKIAA4193; N-acetylglucosamine receptor 1; NAGR1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116655, 4670, 76936 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.57 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P52272, Q62826, Q9D0E1 | |
| HNRNPM | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 491-730 of human HNRNPM (NP_0059592). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |