missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ hnRNP M Monoclonal Antibody (5G6C11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538577
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This antibody has been tested in indirect ELISA.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs. This protein also constitutes a monomer of the N-acetylglucosamine-specific receptor which is postulated to trigger selective recycling of immature GlcNAc-bearing thyroglobulin molecules. Alternative splicing results in multiple transcript variants.
Tekniske data
| hnRNP M | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P52272, Q62826, Q9D0E1 | |
| HNRNPM | |
| Purified recombinant fragment of human NAGR1 (aa: 17-161) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5G6C11 | |
| Unconjugated | |
| HNRNPM | |
| 2610023M21Rik; AA409009; CEA receptor; CEAR; heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; HNRNPM; HNRNPM4; Hnrpm; Hnrpm4; HTGR1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein M; M4 protein; mKIAA4193; N-acetylglucosamine receptor 1; NAGR1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 116655, 4670, 76936 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Spot an opportunity for improvement?Share a Content Correction