missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-6/ONECUT1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99314AF532
Additional Details : Weight : 0.00970kg
Description
HNF-6/ONECUT1 Polyclonal specifically detects HNF-6/ONECUT1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HNF-6/ONECUT1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
3175 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human HNF-6/ONECUT1. | |
0.1 mL | |
Primary | |
Human | |
Purified |