missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
HMGN3/TRIP7 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-59839JF646
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
HMGN3/TRIP7 Polyclonal antibody specifically detects HMGN3/TRIP7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Tekniske data
| HMGN3/TRIP7 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp686E20226, high mobility group nucleosomal binding domain 3, high mobility group nucleosome-binding domain-containing protein 3, high-mobility group protein HMGN3, PNAS-25, thyroid hormone receptor interacting protein 7, thyroid hormone receptor interactor 7, Thyroid receptor-interacting protein 7, TR-interacting protein 7, TRIP-7, TRIP7FLJ42187 | |
| The immunogen recognized by this antibody maps to a region between residue 30 and 60 of human high mobility group nucleosomal binding domain 3 using the numbering given in entry NP_004233.1 (GeneID 9324). | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 9324 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion