missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
HLA DRA Antibody (19-26.1 (same as MB-26.1)), DyLight 405, Novus Biologicals™
Shop alle Bio Techne produkterBeskrivelse
HLA DRA Monoclonal antibody specifically detects HLA DRA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Tekniske data
Tekniske data
| Antigen | HLA DRA |
| Applikationer | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | 19-26.1 (same as MB-26.1) |
| Konjugeret | DyLight 405 |
| Fortynding | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulering | 50mM Sodium Borate |
| Gene Alias | FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW |
| Værtsarter | Mouse |
| Immunogen | Raji cells |
| Vis mere |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?