missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
HLA-DOA 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Description
Western Blotting
Specifications
Specifications
| Cellelinje | 293T |
| Til brug med (applikation) | Western Blot |
| Oversigt | HLA-DOA 293T Cell Transient Overexpression Lysate(Denatured) |
| Genadgangsnr. | BC013183 |
| Molekylvægt (g/mol) | 27.61 |
| pH-område | 6.8 |
| Plasmid | pCMV-HLA-DOA full-length |
| Kvalitetskontrol test | Transient overexpression cell Lysate was tested with Anti-HLA-DOA antibody (H00003111-B01) by western blotting |
| Mængde | 100 μL |
| Opbevaringsbuffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Safety and Handling
Product Identifier
- 100UL HLA-DOA 293T Cell Transient Overexpression L
- Advarsel
- Hudsensibilisering Kategori 1
- H317-Kan forårsage allergisk hudreaktion.
- P261-Undgå indånding af pulver/røg/gas/tåge/damp/spray.
- P272-Tilsmudset arbejdstøj bør ikke fjernes fra arbejdspladsen.
- P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
- MIXTURE LIST-Contain: Mercaptoethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?