missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
HIV-1 Gag p24 Antibody (8G9), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41336MFV450
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA
Tekniske data
HIV-1 Gag p24 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Antibody was raised against a recombinant full-length HIV-1 Gag p24 protein. Amino Acid Squence: pivqniqgqmvhqaisprtlnawvkvveekafspevipmfsalsegatpqdlntmlntvgghqaamqmlketineeaaewdrvhpvhagpiapgqmreprgsdiagttstlqeqigwmtnnppipvgeiykrwiilglnkivrmysptsildirqgpkepfrdyvdrfyktlraeqasqevknwmtetllvqnanpdcktilkalgpaatleemmtacqgvggpghkarvla | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
155030 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
8G9 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
HIV-1 Gag p24 Antibody (8G9), mFluor Violet 450 SE, Novus Biologicals™ > 0.1 mL; mFluor Violet 450 SE
Ser du en mulighed for forbedring?Del en indholdskorrektion