Få mere at vide
Cytiva HiTrap IgSelect Media
Artikelnummer. 11380282 Shop alle Cytiva produkter
Sengevolumen (metrisk):
5 mL
Pakningsstørrelse:
Hver
Artikelnummer. 11380282
Brand: Cytiva 28411312
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
IgSelect is an affinity medium designed for the purification of human IgG from different sources
IgSelect is an affinity medium designed for the purification of human IgG from different sources. The product is a complement to other affinity media with immobilised protein A, such as MabSelect, since it is specific for human IgG and also binds IgG3.The ligand, a 14 kD single chain antibody fragmen, t has been designed for excellent selectivity towards the Fc domain of human IgG, enabling high purity in one step. The IgSelect has affinity for all subclasses of human IgG, making it an excellent complement to protein A based MabSelect media, which has no affinity for IgG3.The ability to bind all subclasses of IgG makes IgSelect an excellent choice for purification of polyclonal human IgG, such as in plasma applications.
  • High purity in one step for human IgG
  • Binds to all subclasses of human IgG, including IgG3
  • Specific to human IgG, no cross reaction with IgG from other species
  • Rigid base matrix allows for high flow rates
  • Suitable for purification of polyclonal human IgG, transgenic IgG, and monoclonal IgG3, IgG4
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 1 x 5 mL
Stabilitet Stable in all commonly used aqueous buffers
Til brug med (applikation) Suitable Purification of Polyclonal Human IgG, Transgenic IgG, and Monoclonal IgG3, IgG4
Opbevaringskrav 4 to 8 deg.C, 20% Ethanol
Bindende Kapacitet 17 mg Human IgG/ml
Produkttype HiTrap IgSelect Media
Partikelstørrelse 75 μm
Materiale Polypropylene
Maks. Tryk 300 kPa
Matrix Highly Cross-linked Agarose
pH-område 2 to 11
Sengevolumen (metrisk) 5 mL
Ligand type 14 kD Recombinant Protein