missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Histone H3.3 Rabbit anti-Human, Mouse, Rat, Clone: 5C5B8, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16489-100UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Histone H3.3 Monoclonal antibody specifically detects Histone H3.3 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunohistochemistry (Paraffin)
Tekniske data
| Histone H3.3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5C5B8 | |
| Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| H3.3A, H3F3, H3F3A, Histone H3.3, Histone H3.3A, Histone H3.3B, MGC87782, MGC87783 | |
| A synthetic peptide corresponding to a sequence within amino acids 57-136 of human Histone H3.3 (P84243). | |
| 100 μg | |
| Chromatin Research, Epigenetics | |
| 3020 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion