missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Histone H2B Antibody (RM230), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26043CL1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Histone H2B Monoclonal antibody specifically detects Histone H2B in Vertebrate samples. It is validated for ELISA,Western Blot,Multiplex Immunoassay,Immunocytochemistry/Immunofluorescence
Tekniske data
| Histone H2B | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| A peptide corresponding to the C-terminus of Human Histone H2B | |
| 0.1 mL | |
| Chromatin Research, Epigenetics | |
| 440689 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot, Multiplex, Immunocytochemistry/Immunofluorescence | |
| RM230 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Vertebrates | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion