Få mere at vide
Histapyrrodine Hydrochloride, LoGiCal
Beskrivelse
Histapyrrodine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Histapyrrodine Hydrochloride |
| CAS | 6113-17-3 |
| Analyt- eller komponentnavne | Histapyrrodine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H24 N2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | N-Phenyl-N-(phenylmethyl)-1-pyrrolidineethanamine hydrochloride (1:1), N-Phenyl-N-(phenylmethyl)-1-pyrrolidineethanamine monohydrochloride, 1-(2-N-Benzylanilinoethyl)pyrrolidine hydrochloride, 1-[2-(N-Benzylanilino)ethyl]pyrrolidine monohydrochloride, 2-(1-Pyrrolidyl)ethylphenylbenzylamine hydrochloride, Calcistin, Domistan, Histapyrrodine hydrochloride, Luvistin, monohydrochloride, N-Benzyl-N-phenyl-2-pyrrolidinoethylamine hydrochloride |
| InChI formel | InChI=1 S/C19H24N2.ClH/c1-3-9-18(10-4-1)17-21(19-11-5-2-6-12-19)16-15-20-13-7-8-14-20;/h1-6,9-12 H,7-8,13-17H2;1 H |
| SMIL | Cl.C(CN1CCCC1)N(Cc2ccccc2)c3ccccc3 |
| IUPAC navn | N-benzyl-N-(2-pyrrolidin-1-ylethyl)aniline;hydrochloride |
| Vis mere |