Learn More
Alfa Aesar™ Histamine
Brand: Alfa Aesar™ J61727.06
Additional Details : CAS Number : 51-45-6 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 5g |
---|
Description
ApplicationsHistamine inhibits the synthesis of IL-2 and γ-IFN in peripheral blood mononuclear cells and lipopolysaccharide-induced synthesis of TNF-α in monocytes via H2 receptor activation. It is a powerful stimulant of gastric secretion, a constrictor of bronchial smooth muscle, a vasodilator, and also a centrally acting neurotransmitter.
Solubility
Soluble in water, alcohol, and hot chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C Hygroscopic. Handle and store under inert gas. Air sensitive.
Chemical Identifiers
51-45-6 | |
111.148 | |
NTYJJOPFIAHURM-UHFFFAOYSA-N | |
774 | |
2-(1H-imidazol-5-yl)ethanamine |
C5H9N3 | |
MFCD00005210 | |
histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine | |
CHEBI:18295 | |
C1=C(NC=N1)CCN |
Specifications
Histamine | |
5g | |
C5H9N3 | |
UN2811 | |
14,4719 | |
Soluble in water,alcohol,and hot chloroform. | |
C1=C(NC=N1)CCN | |
111.148 | |
CHEBI:18295 | |
Powder |
White | |
51-45-6 | |
MFCD00005210 | |
2012 | |
histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine | |
NTYJJOPFIAHURM-UHFFFAOYSA-N | |
2-(1H-imidazol-5-yl)ethanamine | |
774 | |
111.15 |