missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIST1H2AG Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524822
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
Histones are nuclear proteins that play an important role in maintaining the structure of the chromosome in eukaryotes. The nucleosome is made up of around 146bp of DNA, which enfolds around a histone octomer unit comprising of four major histones, H2A, H2B, H3 and H4. An important role of H2A is to ensure proper assembly or functioning of the centrometre. They also play an important role in gene regulation. The protein can be modified by acetylation and phosphorylation.
Specifications
HIST1H2AG | |
Polyclonal | |
Unconjugated | |
H2AC11 | |
H2A histone family, member P; H2A.1; H2A.1b; H2A/d; H2A/p; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; H2AFC; H2AFD; H2AFI; H2AFN; H2AFP; H2AG; H2ak; HIST1H2AG; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; histone 1, H2ag; histone cluster 1, H2ag; histone H2A type 1; histone H2A/e; histone H2A/p; Histone H2A/ptl; pH2A/f | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8329, 8969 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P0C0S8 | |
H2AC11, HIST1H2AG | |
KLH conjugated synthetic peptide between 103-130 amino acids from the C-terminal region of human HIST1H2AL. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ HIST1H2AG Polyclonal Antibody > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction